missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDP2 Protein
A DNA sequence encoding the Homo sapiens (Human) PDP2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8098-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001316857.1 | |
PDP2 Protein | |
Research Use Only | |
PDP2 | |
Recombinant Protein | |
E. coli | |
STEEDDFHLQ LSPEQINEVL RAGETTHKIL DLESRVPNSV LRFESNQLAA NSPVEDRRGV ASCLQTNGLM FGIFDGHGGH ACAQAVSERL FYYVAVSLMS HQTLEHMEGA MESMKPLLPI LHWLKHPGDS IYKDVTSVHL DHLRVYWQEL LDLHMEMGLS IEEALMYSFQ RLDSDISLEI QAPLEDEVTR NLSLQVAFSG ATACMAHVDG IHLHVANAGD CRAILGVQED NGMWSCLPLT RDHNAWNQAE LSRLKREHPE SEDRTIIMED RLLGVLIPCR AFGDVQLKWS KELQRSILER GFNTEALNIY QFTPPHYYTP PYLTAEPEVT YHRLRPQDKF LVLASDGLWD MLSNEDVVRL VVGHLAEADW HKTDLAQRPA NLGLMQSLLL QRKASGLHEA DQNAATRLIR HAIGNNEYGE MEAERLAAML TLPEDLARMY RDDITVTVVY FNSESIGAYY KGG | |
Greater than 90% as determined by SDS-PAGE. |
57546 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |