missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peroxiredoxin 1 / PRDX1 Protein
A DNA sequence encoding the Homo sapiens (Human) Peroxiredoxin 1 / PRDX1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8352-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001189360.1 | |
Peroxiredoxin 1 / PRDX1 Protein | |
Research Use Only | |
PRDX1 | |
Recombinant Protein | |
E. coli | |
MSSGNAKIGH PAPNFKATAV MPDGQFKDIS LSDYKGKYVV FFFYPLDFTF VCPTEIIAFS DRAEEFKKLN CQVIGASVDS HFCHLAWVNT PKKQGGLGPM NIPLVSDPKR TIAQDYGVLK ADEGISFRGL FIIDDKGILR QITVNDLPVG RSVDETLRLV QAFQFTDKHG EVCPAGWKPG SDTIKPDVQK SKEYFSKQK | |
Greater than 90% as determined by SDS-PAGE. |
5052 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |