missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGK1 / PGKA Protein
A DNA sequence encoding the Homo sapiens (Human) PGK1 / PGKA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8305-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000282.1 | |
PGK1 / PGKA Protein | |
Research Use Only | |
PGK1 | |
Recombinant Protein | |
E. coli | |
FNVPMKNNQI TNNQRIKAAV PSIKFCLDNG AKSVVLMSHL GRPDGVPMPD KYSLEPVAVE LKSLLGKDVL FLKDCVGPEV EKACANPAAG SVILLENLRF HVEEEGKGKD ASGNKVKAEP AKIEAFRASL SKLGDVYVND AFGTAHRAHS SMVGVNLPQK AGGFLMKKEL NYFAKALESP ERPFLAILGG AKVADKIQLI NNMLDKVNEM IIGGGMAFTF LKVLNNMEIG TSLFDEEGAK IVKDLMSKAE KNGVKITLPV DFVTADKFDE NAKTGQATVA SGIPAGWMGL DCGPESSKKY AEAVTRAKQI VWNGPVGVFE WEAFARGTKA LMDEVVKATS RGCITIIGGG DTATCCAKWN TEDKVSHVST GGGASLELLE GKVLPGVDAL SNI | |
Greater than 90% as determined by SDS-PAGE. |
5230 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |