missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGLYRP1 Protein
A cDNA sequence encoding the PGLYRP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13033-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PGLYRP1 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PGLYRP1 filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.0. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASQETEDPACCS PIVPRNEWKA LASECAQHLS LPLRYVVVSH TAGSSCNTPA SCQQQARNVQ HYHMKTLGWC DVGYNFLIGE DGLVYEGRGW NFTGAHSGHL WNPMSIGISF MGNYMDRVPT PQAIRAAQGL LACGVAQGAL RSNYVLKGHR DVQRTLSPGN QLYHLIQNWP HYRSP |