missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phosphoserine phosphatase Protein
A DNA sequence encoding the Homo sapiens (Human) Phosphoserine phosphatase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6549-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004568.2 | |
Phosphoserine phosphatase Protein | |
Research Use Only | |
PSPH | |
Recombinant Protein | |
E. coli | |
MVSHSELRKL FYSADAVCFD VDSTVIREEG IDELAKICGV EDAVSEMTRR AMGGAVPFKA ALTERLALIQ PSREQVQRLI AEQPPHLTPG IRELVSRLQE RNVQVFLISG GFRSIVEHVA SKLNIPATNV FANRLKFYFN GEYAGFDETQ PTAESGGKGK VIKLLKEKFH FKKIIMIGDG ATDMEACPPA DAFIGFGGNV IRQQVKDNAK WYITDFVELL GELEE | |
Greater than 80% as determined by SDS-PAGE. |
5723 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |