missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PI3Kb Protein

A cDNA sequence encoding the PI3Kb was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10818-5ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15905268

  • 605.00€ / 5µg

Spedizione stimata: 03-04-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

PI3Kb Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Insect
0.9 mg/ml solution in PBS and 2mM MgCl2.
Greater than 90.0% as determined by SDS Page.
Research Use Only
The specific activity was found to be 3,000 units/mg (1 unit is defined as 1 picomole phosphate transferred to PIP2 per minute).
Human
Untagged
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali