missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIK3R5 Protein
A DNA sequence encoding the Homo sapiens (Human) PIK3R5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7909-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001136105.1 | |
PIK3R5 Protein | |
Research Use Only | |
PIK3R5 | |
Recombinant Protein | |
E. coli | |
MDSGYVEDSE ESSSEWPWRR GSQERRGHRR PGQKFIRIYK LFKSTSQLVL RRDSRSLEGS SDTALPLRRA GSLCSPLDEP VSPPSRAQRS RSLPQPKLGT QLPSWLLAPA SRPQRRRPFL SGDEDPKAST LRVVVFGSDR ISGKVARAYS NLRRLENNRP LLTRFFKLQF FYVPVKRSHG TSPGACPPPR SQTPSPPTDS PRHASPGELG TTPWEESTND ISHYLGMLDP WYERNVLGLM HLPPEVLCQQ SLKAEAQALE GSPTQLPILA DMLLYYCRFA ARPVLLQVYQ TELTFITGEK TTEIFIHSLE LGHSAATRAI KASGPGSKRL GIDGDREAVP LTLQIIYSKG AISGRSRWSN LEKVCTSVNL NKACRKQEEL DSSMEALTLN LTEVVKRQNS KSKKGFNQIS TSQIKVDKVQ IIGSNSCPFA VCLDQDERKI LQSVVRCEVS PCYKPEKSDL SSPPQTPPDL PAQAAPDLCS LLCLPIMTFS GALP | |
Greater than 90% as determined by SDS-PAGE. |
23533 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |