missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLL Protein
A cDNA sequence encoding the POLL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13094-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
POLL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLMHHQKYLQ RFLGGKREKK QKEACSIPGI GKRMAEKIIE ILESGHLRKL DHISESVPVL ELFSNIWGAG TKTAQMWYQQ GFRSLEDIRS QASLTTQQAI GLKHYSDFLE RMPREEATEI EQTVQKAAQA FNSGLLCVAC GSYRRGKATC GDVDVLITHP DGRSHRGIFS RLLDSLRQEG FLTDDLVSQE ENGQQQKYLG VCRLPGPGRR HRRLDIIVVP YSEFACALLY FTGSAHFNRS MRALAKTKGM SLSEHALSTA VVRNTHGCKV GPGRVLPTPT EKDVFRLLGL PYREPAERDW | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POLL solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 30% glycerol. |