missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1CA Protein
A cDNA sequence encoding the PPP1CA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10835-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PPP1CA Protein | |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSDSEKLNLD SIIGRLLEVQ GSRPGKNVQL TENEIRGLCL KSREIFLSQP ILLELEAPLK ICGDIHGQYY DLLRLFEYGG FPPESNYLFL GDYVDRGKQS LETICLLLAY KIKYPENFFL LRGNHECASI NRIYGFYDEC KRRYNIKLWK TFTDCFNCLP IAAIVDEKIF CCHGGLSPDL QSMEQIRRIM RPTDVPDQGL LCDLLWSDPD KDVQGWGEND RGVSFTFGAE VVAKFLHKHD LDLICRAHQV VEDGYEFFAK RQLVTLFSAP NYCGEFDNAG AMMSVDETLM CSFQILKPAD KNKGKYGQFS GLNPGGRPIT PPRNSAKAKK |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Specific activity: >3,000 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme hydrolyzing 1 nmole of p-nitrophenyl phosphate (pNPP) per minute at 37C, pH7.5 using 10mM of substrate. | |
Human | |
His | |
PPP1CA protein solution (0.2 mg/ml) containing 50mM Tris-HCl (pH 8.5),0.2M NaCl, 1mM DTT, 0.1mM PMSF, 1mM MnCl2 and 50% glycerol. |