missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pregnancy zone Protein
A DNA sequence encoding the Homo sapiens (Human) Pregnancy zone protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6563-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Pregnancy zone Protein | |
Research Use Only | |
PZP | |
Recombinant Protein | |
E. coli | |
MKPEAELSVS SVYNLLTVKD LTNFPDNVDQ QEEEQGHCPR PFFIHNGAIY VPLSSNEADI YSFLKGMGLK VFTNSKIRKP KSCSVIPSVS AGAVGQGYYG AGLGVVERPY VPQLGTYNVI PLNNEQSSGP VPETVRSYFP ETWIWELVAV NSSGVAEVGV TVPDTITEWK AGAFCLSEDA GLGISSTASL RAFQPFFVEL TMPYSVIRGE VFTLKATVLN YLPKCIRAEG IEQEKTFSSM TCASGANVSE QLSLKLPSNV VKESARASFS VLGDILGSAM QNIQNLLQMP YGCGEQNMVL FAPNIYVLNY LNETQQLTQE IKAKAVGYLI TGYQRQLNYK HQDGSYSTFG | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |