missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable imidazolonepropionase Protein
A DNA sequence encoding the Homo sapiens (Human) Probable imidazolonepropionase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7963-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_689648.2 | |
Probable imidazolonepropionase Protein | |
Research Use Only | |
AMDHD1 | |
Recombinant Protein | |
E. coli | |
MASGHSLLLE NAQQVVLVCA RGERFLARDA LRSLAVLEGA SLVVGKDGFI KAIGPADVIQ RQFSGETFEE IIDCSGKCIL PGLVDAHTHP VWAGERVHEF AMKLAGATYM EIHQAGGGIH FTVERTRQAT EEELFRSLQQ RLQCMMRAGT TLVECKSGYG LDLETELKML RVIERARREL DIGISATYCG AHSVPKGKTA TEAADDIINN HLPKLKELGR NGEIHVDNID VFCEKGVFDL DSTRRILQRG KDIGLQINFH GDELHPMKAA ELGAELGAQA ISHLEEVSDE GIVAMATARC SAILLPTTAY MLRLKQPRAR KMLDEGVIVA LGSDFNPNAY CFSMPMVMHL ACVNMRMSMP EALAAATINA AYALGKSHTH GSLEVGKQGD LIIINSSRWE HLIYQFGGHH ELIEYVIAKG KLIYKT | |
Greater than 90% as determined by SDS-PAGE. |
144193 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |