missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-methionine sulfoxide oxidase MICAL2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-methionine sulfoxide oxidase MICAL2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9273-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001269592.1 | |
Protein-methionine sulfoxide oxidase MICAL2 Protein | |
Research Use Only | |
MICAL2 | |
Recombinant Protein | |
Yeast | |
MGENEDEKQA QAGQVFENFV QASTCKGTLQ AFNILTRHLD LDPLDHRNFY SKLKSKVTTW KAKALWYKLD KRGSHKEYKR GKSCTNTKCL IVGGGPCGLR TAIELAYLGA KVVVVEKRDS FSRNNVLHLW PFTIHDLRGL GAKKFYGKFC AGSIDHISIR QLQLILFKVA LMLGVEIHVN VEFVKVLEPP EDQENQKIGW RAEFLPTDHS LSEFEFDVII GADGRRNTLE GFRRKEFRGK LAIAITANFI NRNSTAEAKV EEISGVAFIF NQKFFQDLKE ETGIDLENIV YYKDCTHYFV MTAKKQSLLD KGVIINDYID TEMLLCAENV NQDNLLSYAR EAADFATNYQ LPSLDFAMNH YGQPDVAMFD FTCMYASENA ALVRERQAHQ LLVALVGDSL LEPFWPMGTG CARGFLAAFD TAWMVKSWNQ GTPPLELLAE RESLYRLLPQ TTPENINKNF EQYTLDPGTR YPNLNSHCVR PHQVKHLYIT KELEH | |
Greater than 90% as determined by SDS-PAGE. |
9645 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |