missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Wnt-3a Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-3a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8838-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_149122.1 | |
Protein Wnt-3a Protein | |
Research Use Only | |
Wnt3a | |
Recombinant Protein | |
E. coli | |
SYPIWWSLAV GPQYSSLGSQ PILCASIPGL VPKQLRFCRN YVEIMPSVAE GIKIGIQECQ HQFRGRRWNC TTVHDSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGTAAICGC SSRHQGSPGK GWKWGGCSED IEFGGMVSRE FADARENRPD ARSAMNRHNN EAGRQAIASH MHLKCKCHGL SGSCEVKTCW WSQPDFRAIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRP RYTYFKVPTE RDLVYYEASP NFCEPNPETG SFGTRDRTCN VSSHGIDGCD LLCCGRGHNA RAERRREKCR CVFHWCCYVS CQECTRVYDV HTC | |
Greater than 90% as determined by SDS-PAGE. |
89780 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |