missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTGES3 Protein
A cDNA sequence encoding the PTGES3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13201-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PTGES3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MQPASAKWYD RRDYVFIEFC VEDSKDVNVN FEKSKLTFSC LGGSDNFKHL NEIDLFHCID PNDSKHKRTD RSILCCLRKG ESGQSWPRLT KERAKLNWLS VDFNNWKDWE DDSDEDMSNF DRFSEMMNNM GGDEDVDLPE VDGADDDSQD SDDEKMPDLE | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The PTGES3 protein solution contains 20mM Tris-HCl, pH 8.0, 1mM DTT and 10% Glycerol. |