missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTPN1 Protein

A cDNA sequence encoding the PTPN1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10856-50ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15945378

  • 181.00€ / 50µg

Spedizione stimata: 02-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

PTPN1 Protein
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MEMEKEFEQI DKSGSWAAIY QDIRHEASDF PCRVAKLPKN KNRNRYRDVS PFDHSRIKLHQEDNDYINAS LIKMEEAQRS YILTQGPLPN TCGHFWEMVW EQKSRGVVML NRVMEKGSLKCAQYWPQKEE KEMIFEDTNL KLTLISEDIK SYYTVRQLEL ENLTTQETRE ILHFHYTTWPDFGVPESPAS FLNFLFKVRE SGSLSPEHGP VVVHCSAGIG RSGTFCLADT CLLLMDKRKDPSSVDIKKVL LEMRKFRMGL IQTADQLRFS YLAVIEGAKF IMGDSSVQDQ WKELSHEDLE PPPEHIPPPPRPPKRILEPHN
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 10,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C.
Human
Untagged
The protein (1 mg/ml) contains 25mM Tris-HCl, pH 7.5, 2mM beta-mercaptoethanol, 1mM EDTA, 1mM DTT and 20% Glycerol.
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali