missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-8A Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-8A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6575-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005361.2 | |
Ras-related protein Rab-8A Protein | |
Research Use Only | |
RAB8A | |
Recombinant Protein | |
E. coli | |
KTYDYLFKLL LIGDSGVGKT CVLFRFSEDA FNSTFISTIG IDFKIRTIEL DGKRIKLQIW DTAGQERFRT ITTAYYRGAM GIMLVYDITN EKSFDNIRNW IRNIEEHASA DVEKMILGNK CDVNDKRQVS KERGEKLALD YGIKFMETSA KANINVENAF FTLARDIKAK MDKKLEGNSP QGSNQGVKIT P | |
Greater than 90% as determined by SDS-PAGE. |
4218 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |