missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBMY1A1 Protein
A DNA sequence encoding the Homo sapiens (Human) RBMY1A1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7035-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001307873.1 | |
RBMY1A1 Protein | |
Research Use Only | |
RBMY1A1 | |
Recombinant Protein | |
E. coli | |
MVEADHPGKL FIGGLNRETN EKMLKAVFGK HGPISEVLLI KDRTSKSRGF AFITFENPAD AKNAAKDMNG KSLHGKAIKV EQAKKPSFQS GGRRRPPASS RNRSPSGSLR SARGSRGGTR GWLPSHEGHL DDGGYTPDLK MSYSRGLIPV KRGPSSRSGG PPPKKSAPSA VARSNSWMGS QGPMSQRREN YGVPPRRATI SSWRNDRMST RHDGYATNDG NHPSCQETRD YAPPSRGYAY RDNGHSNRDE HSSRGYRNHR SSRETRDYAP PSRGHAYRDY GHSRRDESYS RGYRNRRSSR ETREYAPPSR GHGYRDYGHS RRHESYSRGY RNHPSSRETR DYAPPHRDYA YRDYGHSSWD EHSSRGYSYH DGYGEALGRD HSEHLSGSSY RDALQRYGTS HGAPPARGPR MSYGGSTCHA YSNTRDRYGR SWESYSSCGD FHYCDREHVC RKDQRNPPS | |
Greater than 90% as determined by SDS-PAGE. |
378949 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |