missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RFXANK Protein
A cDNA sequence encoding the RFXANK was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13304-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
8625 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMELTQPA EDLIQTQQTP ASELGDPEDP GEEAADGSDT VVLSLFPCTP EPVNPEPDAS VSSPQGSSLK HSTTLTNRQR GNEVSALPAT LDCDNLVNKP DERGFTPLIW ASAFGEIETV RFLLEWGADP HILAKERESA LSLASTGGYT DIVGLLLERD VDINIYDWNG GTPLLYAVRG NHVKCVEALL ARGADLTTEA DSGYTPMDLA VALGYRKVQQ VIENHILKLF QSNLVPADPE | |
Greater than 85% as determined by SDS-PAGE. |
RFXANK Protein | |
Research Use Only | |
RFXANK | |
Human | |
His | |
The RFXANK solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |