missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Semenogelin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Semenogelin-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9386-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002999.1 | |
Semenogelin-2 Protein | |
Research Use Only | |
SEMG2 | |
Recombinant Protein | |
Yeast | |
QKGGSKGQLP SGSSQFPHGQ KGQHYFGQKD QQHTKSKGSF SIQHTYHVDI NDHDWTRKSQ QYDLNALHKA TKSKQHLGGS QQLLNYKQEG RDHDKSKGHF HMIVIHHKGG QAHHGTQNPS QDQGNSPSGK GLSSQCSNTE KRLWVHGLSK EQASASGAQK GRTQGGSQSS YVLQTEELVV NKQQRETKNS HQNKGHYQNV VDVREEHSSK LQTSLHPAHQ DRLQHGPKDI FTTQDELLVY NKNQHQTKNL SQDQEHGRKA HKISYPSSRT EERQLHHGEK SVQKDVSKGS ISIQTEEKIH GKSQNQVTIH SQDQEHGHKE NKISYQSSST EERHLNCGEK GIQKGVSKGS ISIQTEEQIH GKSQNQVRIP SQAQEYGHKE NKISYQSSST EERRLNSGEK DVQKGVSKGS ISIQTEEKIH GKSQNQVTIP SQDQEHGHKE NKMSYQSSST EERRLNYGGK STQKDVSQSS ISFQIEKLVE GKSQIQTPNP NQDQWSGQNA KGKSGQSADS KQDLLSHEQK GRYKQESSES HNIVITEHEV AQDDHLTQQY NEDRNPIST | |
Greater than 90% as determined by SDS-PAGE. |
6407 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |