missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINH1 Protein
A cDNA sequence encoding the SERPINH1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13469-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
SERPINH1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAEVKKPAA AAAPGTAEKL SPKAATLAER SAGLAFSLYQ AMAKDQAVEN ILVSPVVVAS SLGLVSLGGK ATTASQAKAV LSAEQLRDEE VHAGLGELLR SLSNSTARNV TWKLGSRLYG PSSVSFADDF VRSSKQHYNC EHSKINFRDK RSALQSINEW AAQTTDGKLP EVTKDVERTD GALLVNAMFF KPHWDEKFHH KMVDNRGFMV TRSYTVGVMM MHRTGLYNYY DDEKEKLQIV EMPLAHKLSS LIILMPHHVE PLERLEKLLT KEQLKIWMGK MQKKAVAISL PKGVVEVTHD LQKHLAGLGL TEAIDKNKAD LSRMSGKKDL YLASVFHATA FELDTDGNPF DQDIYGREEL RSPKLFYADH PFIFLVRDTQ SGSLLFIGRL VRPKGDKMRD EL | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SERPINH1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |