missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SGCD Protein
A cDNA sequence encoding the SGCD was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13482-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
SGCD Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH RSHMKVMNFT IDGMGNLRIT EKGLKLEGDS EFLQPLYAKE IQSRPGNALY FKSARNVTVN ILNDQTKVLT QLITGPKAVE AYGKKFEVKT VSGKLLFSAD NNEVVVGAER LRVLGAEGTV FPKSIETPNV RADPFKELRL ESPTRSLVME APKGVEINAE AGNMEATCRT ELRLESKDGE IKLDAAKIRL PRLPHGSYTP TGTRQKVFEI CVCANGRLFL SQAGAGSTCQ INTSVCL | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SGCD protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |