missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sialic acid-binding Ig-like lectin 15 Protein
A DNA sequence encoding the Homo sapiens (Human) Sialic acid-binding Ig-like lectin 15, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7774-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_998767.1 | |
Sialic acid-binding Ig-like lectin 15 Protein | |
Research Use Only | |
SIGLEC15 | |
Recombinant Protein | |
E. coli | |
FVRTKIDTTE NLLNTEVHSS PAQRWSMQVP PEVSAEAGDA AVLPCTFTHP HRHYDGPLTA IWRAGEPYAG PQVFRCAAAR GSELCQTALS LHGRFRLLGN PRRNDLSLRV ERLALADDRR YFCRVEFAGD VHDRYESRHG VRLHVTAAPR IVNISVLPSP AHAFRALCTA EGEPPPALAW SGPALGNSLA AVRSPREGHG HLVTAELPAL THDGRYTCTA ANSLGRSEAS VYLFRFHGAS GAST | |
Greater than 90% as determined by SDS-PAGE. |
284266 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |