Learn More
enQuireBio™ Recombinant Human STMN1 Protein
A cDNA sequence encoding the STMN1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13620-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
STMN1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASSDIQVKE LEKRASGQAF ELILSPRSKE SVPEFPLSPP KKKDLSLEEI QKKLEAAEER RKSHEAEVLK QLAEKREHEK EVLQKAIEEN NNFSKMAEEK LTHKMEANKE NREAQMAAKL ERLREKDKHI EEVRKNKESK DPADETEAD | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The STMN1 protein solution contains 20mM Tris-HCl, pH 8 and 10% Glycerol. |