missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAC3 Protein
A cDNA sequence encoding the TAC3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13680-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
TAC3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH QSFGAVCKEP QEEVVPGGGR SKRDPDLYQL LQRLFKSHSS LEGLLKALSQ ASTDPKESTS PEKRDMHDFF VGLMGKRSVQ PDSPTDVNQE NVPSFGILKY PPRAE | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TAC3 solution contains 20mM Tris-HCl buffer (pH 8.0) and 20% glycerol. |