missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMP1 Protein
A cDNA sequence encoding the TIMP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10881-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
TIMP1 Protein | |
Research Use Only | |
The IC50 of 2.5-4 nM is measured by its ability to inhibit recombinant human MMP-2 cleavage of the colorimetric peptide substrate, Mca-PLGL-DpaAR-NH2. | |
Human | |
His | |
The TIMP1 protein was lyophilized after extensive dialysis against PBS. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
MAPFEPLASG ILLLLWLIAP SRACTCVPPH PQTAFCNSDL VIRAKFVGTP EVNQTTLYQR YEIKMTKMYK GFQALGDAAD IRFVYTPAME SVCGYFHRSH NRSEEFLIAG KLQDGLLHIT TCSFVAPWNS LSLAQRRGFT KTYTVGCEEC TVFPCLSIPC KLQSGTHCLW TDQLLQGSEK GFQSRHLACL PREPGLCTWQ SLRSQIA | |
Greater than 95.0% as determined by SDS-PAGE. |