missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Titin Protein
A DNA sequence encoding the Homo sapiens (Human) Titin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8823-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001243779.1 | |
Titin Protein | |
Research Use Only | |
TTN | |
Recombinant Protein | |
E. coli | |
VGSSICSARV TLREPPSFIK KIESTSSLRG GTAAFQATLK GSLPITVTWL KDSDEITEDD NIRMTFENNV ASLYLSGIEV KHDGKYVCQA KNDAGIQRCS ALLSVKEPAT ITEEAVSIDV TQGDPATLQV KFSGTKEITA KWFKDGQELT LGSKYKISVT DTVSILKIIS TEKKDSGEYT FEVQNDVGRS SCKARINVLD LIIPPSF | |
Greater than 90% as determined by SDS-PAGE. |
7273 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |