missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TL1A / TNFSF15 Protein
A DNA sequence encoding the Human TL1A / TNFSF15, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10372-ec-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001191273.1 | |
TL1A / TNFSF15 Protein | |
Research Use Only | |
TNFSF15 | |
The ED50 as determined by its ability to induce apoptosis using human TF-1 cells is less than 20 ng/ml, corresponding to a specific activity of > 5.0 M 104 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, with 0.02% Tween-20, lyophilized |
9966 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LKGQEFAPSH QQVYAPLRAD GDKPRAHLTV VRQTPTQHFK NQFPALHWEH ELGLAFTKNR MNYTNKFLLI PESGDYFIYS QVTFRGMTSE CSEIRQAGRP NKPDSITVVI TKVTDSYPEP TQLLMGTKSV CEVGSNWFQP IYLGAMFSLQ EGDKLMVNVS DISLVDYTKE DKTFFGAFLL | |
Greater than 97% as determined by SDS-PAGE and HPLC. |