missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFR1 / CD120a / TNFRSF1A Protein
A DNA sequence encoding the Human TNFR1 / CD120a / TNFRSF1A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10374-ec-250ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001056.1 | |
TNFR1 / CD120a / TNFRSF1A Protein | |
Research Use Only | |
TNFRSF1A | |
The ED50 as determined by its ability to inhibit the TNF-alpha mediated cytotoxicity in the L-929 cells is less than 0.05 ug/ml, corresponding to a specific activity of > 2 M 104 IU/mg in the presence of 0.25 ng/mL of rHuTNF-alpha. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
7132 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
IYPSGVIGLV PHLGDREKRD SVCPQGKYIH PQNNSICCTK CHKGTYLYND CPGPGQDTDC RECESGSFTA SENHLRHCLS CSKCRKEMGQ VEISSCTVDR DTVCGCRKNQ YRHYWSENLF QCFNCSLCLN GTVHLSCQEK QNTVCTCHAG FFLRENECVS CSNCKKSLEC TKLCLPQIEN VKGTEDSGTT | |
Greater than 97% as determined by SDS-PAGE and HPLC. |