missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF10A Protein
A cDNA sequence encoding the TNFRSF10A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10918-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
8797 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
TNFRSF10A | |
Human | |
His | |
TNFRSF10A protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
TNFRSF10A Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Insect | |
ASGTEAAAAT PSKVWGSSAG RIEPRGGGRG ALPTSMGQHG PSARARAGRA PGPRPAREAS PRLRVHKTFK FVVVGVLLQV VPSSAATIKL HDQSIGTQQW EHSPLGELCP PGSHRSEHPG ACNRCTEGVG YTNASNNLFA CLPCTACKSD EEERSPCTTT RNTACQCKPG TFRNDNSAEM CRKCSRGCPR GMVKVKDCTP WSDIECVHKE SGNGHNLEHH HHHH |