missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFSF10 / TRAIL / APO-2L Protein
Recombinant Protein
Marca: enQuireBio™ QP5244-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001177871.1 | |
TNFSF10 / TRAIL / APO-2L Protein | |
Research Use Only | |
TNFSF10 | |
Recombinant Protein | |
E. coli | |
MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG | |
Greater than 90 as determined by SDS-PAGE. |
8743 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human TNFSF10 / TRAIL / APO-2L induces RPMI-8226 cell proliferation (typical ED50 is ? 15 ng/ml). | |
Human | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |