missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TPI1 Protein
A cDNA sequence encoding the TPI1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10891-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
7167 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
TPI1 | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAPSRKFFVG GNWKMNGRKQ SLGELIGTLN AAKVPADTEV VCAPPTAYID FARQKLDPKI AVAAQNCYKV TNGAFTGEIS PGMIKDCGAT WVVLGHSERR HVFGESDELI GQKVAHALAE GLGVIACIGE KLDEREAGIT EKVVFEQTKV IADNVKDWSK VVLAYEPVWA IGTGKTATPQ QAQEVHEKLR GWLKSNVSDA VAQSTRIIYG GSVTGATCKE LASQPDVDGF LVGGASLKPE FVDIINAKQ |
TPI1 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 3000 units/mg, in which one unit will convert 1.0 umole of D-glyceraldehyde-3-phosphate to dihydroxyacetone phosphate per minute at pH 7.5 at 25C. | |
Human | |
His | |
The TPI1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |