missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UGT8 Protein
A cDNA sequence encoding the UGT8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13884-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
UGT8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAKIIIVP PIMFESHMYI FKTLASALHE RGHHTVFLLS EGRDIAPSNH YSLQRYPGIF NSTTSDAFLQ SKMRNIFSGR LTAIELFDIL DHYTKNCDLM VGNHALIQGL KKEKFDLLLV DPNDMCGFVI AHLLGVKYAV FSTGLWYPAE VGAPAPLAYV PEFNSLLTDR MNLLQRMKNT GVYLISRLGV SFLVLPKYER IMQKYNLLPE KSMYDLVHGS SLWMLCTDVA LEFPRPTLPN VVYVGGILTK PASPLPEDLQ RWVNGANEHG FVLVSFGAGV KYLSEDIANK LAGALGRLPQ KVIWRFSGPK PKNLGNNTKL IEWLPQNDLL GHSKIKAFLS HGGLNSIFET MYHGVPVVGI PLFGDHYDTM TRVQAKGMGI LLEWKTVTEK ELYEALVKVI NNPSYRQRAQ KLSEIHKDQP GHPVNRTIYW IDYIIRHNGA HHLRAAVHQI SFCQYFLLDI AFVLLLGAAL LYFLLSWVTK FIYRKIKSLW SRNKHSTVNG HYHNGILNGK YKRNGHIKHE KKVK | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UGT8 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |