missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPK3A Protein
A cDNA sequence encoding the UPK3A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13898-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
UPK3A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMVNLQP QLASVTFATN NPTLTTVALE KPLCMFDSKE ALTGTHEVYL YVLVDSAISR NASVQDSTNT PLGSTFLQTE GGRTGPYKAV AFDLIPCSDL PSLDAIGDVS KASQILNAYL VRVGANGTCL WDPNFQGLCN PPLSAATEYR FKYVLVNMST GLVEDQTLWS DPIRTNQLTP YSTIDTWPGR RSGG | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UPK3A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, 2mM DTT and 20% glycerol. |