missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VGLL3 Protein
A DNA sequence encoding the Homo sapiens (Human) VGLL3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9465-ye-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001307422.1 | |
VGLL3 Protein | |
Research Use Only | |
VGLL3 | |
Recombinant Protein | |
Yeast | |
MSCAEVMYHP QPYGASQYLP NPMAATTCPT AYYQPAPQPG QQKKLAVFSK MQDSLEVTLP SKQEEEDEEE EEEEKDQPAE MEYLNSRCVL FTYFQGDIGS VVDEHFSRAL GQAITLHPES AISKSKMGLT PLWRDSSALS SQRNSFPTSF WTSSYQPPPA PCLGGVHPDF QVTGPPGTFS AADPSPWPGH NLHQTGPAPP PAVSESWPYP LTSQVSPSYS HMHDVYMRHH HPHAHMHHRH RHHHHHHHPP AGSALDPSYG PLLMPSVHAA RIPAPQCDIT KTEPTTVTSA TSAWAGAFHG TVDIVPSVGF DTGWSAMARS | |
Greater than 90% as determined by SDS-PAGE. |
389136 | |
1 mg | |
Not determined. | |
Full-Length (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |