missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Ohio Protein
A cDNA sequence encoding the Influenza-B Ohio was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12441-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Influenza-B Ohio Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant B/Ohio/01/05 solution contains 30mM Sodium phosphate, pH 7.4 and 50mM NaCl. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |