missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque FLT3L / Flt3 ligand Protein
A DNA sequence encoding the Rhesus Macaque FLT3L / Flt3 ligand, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10447-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
XP_014980066.1 | |
FLT3L / Flt3 ligand Protein | |
Research Use Only | |
FLT3LG | |
The ED50 as determined by a cell proliferation assay using human AML5 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
719239 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVPSNLQD EELCGALWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQHP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPRSPGALEA TALTAPQRP | |
Greater than 97% as determined by SDS-PAGE and HPLC. |