missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque IL4 / Interleukin-4 Protein
A DNA sequence encoding the Rhesus Macaque IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10337-ec-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
IL4 / Interleukin-4 Protein | |
Research Use Only | |
IL4 | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Macaque | |
Untagged | |
0.2 um filtered 2 M PBS, pH 7.4, 5 % trehalose, lyophilized |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
HNCHIALREI IETLNSLTEQ KTLCTKLTIT DILAASKNTT EKETFCRAAT VLRQFYSHHE KDTRCLGATA QQFHRHKQLI RFLKRLDRNL WGLAGLNSCP VKEANQSTLE DFLERLKTIM REKYSKCSS | |
Greater than 96% as determined by SDS-PAGE and HPLC. |