missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse 2-oxoglutarate dehydrogenase, mitochondrial Protein
A DNA sequence encoding the Mus musculus (Mouse) 2-oxoglutarate dehydrogenase, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7717-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001239211.1 | |
2-oxoglutarate dehydrogenase, mitochondrial Protein | |
Research Use Only | |
OGDH | |
Recombinant Protein | |
E. coli | |
HLWPNQAPYT VCNSSLSEYG VLGFELGFAM ASPNALVLWE AQFGDFNNMA QCIIDQFICP GQAKWVRQNG IVLLLPHGME GMGPEHSSAR PERFLQMCND DPDVLPDLQE ENFDINQLYD CNWIVVNCST PGNFFHVLRR QILLPFRKPL IVFTPKSLLR HPEARTSFDE MLPGTHFQRV IPENGPAAQD PHKVKRLLFC TGKVYYDLTR ERKARNMEEE VAITRIEQLS PFPFDLLLKE AQKYPNAELA WCQEEHKNQG YYDYVKPRLR TTIDRAKPVW YAGRDPAAAP ATGNKKTHLT ELQRFLDTAF DLDAFKKFS | |
Greater than 90% as determined by SDS-PAGE. |
18293 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |