missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Acyl-protein thioesterase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Acyl-protein thioesterase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6328-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_032892.1 | |
Acyl-protein thioesterase 1 Protein | |
Research Use Only | |
Lypla1 | |
Recombinant Protein | |
E. coli | |
MCGNNMSAPM PAVVPAARKA TAAVIFLHGL GDTGHGWAEA FAGIKSPHIK YICPHAPVMP VTLNMNMAMP SWFDIVGLSP DSQEDESGIK QAAETVKALI DQEVKNGIPS NRIILGGFSQ GGALSLYTAL TTQQKLAGVT ALSCWLPLRA SFSQGPINSA NRDISVLQCH GDCDPLVPLM FGSLTVERLK ALINPANVTF KIYEGMMHSS CQQEMMDVKH FIDKLLPPID | |
Greater than 90% as determined by SDS-PAGE. |
18777 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |