Learn More
enQuireBio™ Recombinant Mouse bFGF / FGF2 Protein
Recombinant Protein
Marca: enQuireBio™ QP5413-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_032032.1 | |
bFGF / FGF2 Protein | |
Research Use Only | |
FGF2 | |
Recombinant Protein | |
E. coli | |
MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS | |
Greater than 95% as determined by SDS-PAGE. |
14173 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse bFGF / FGF2 induces 3T3 cell proliferation (typical ED50 is ? 0.5 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |