missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CALR / Calreticulin Protein
A DNA sequence encoding the Mus musculus (Mouse) CALR / Calreticulin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5750-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_031617.1 | |
CALR / Calreticulin Protein | |
Research Use Only | |
Calr | |
Recombinant Protein | |
E. coli | |
DPAIYFKEQF LDGDAWTNRW VESKHKSDFG KFVLSSGKFY GDLEKDKGLQ TSQDARFYAL SAKFEPFSNK GQTLVVQFTV KHEQNIDCGG GYVKLFPSGL DQKDMHGDSE YNIMFGPDIC GPGTKKVHVI FNYKGKNVLI NKDIRCKDDE FTHLYTLIVR PDNTYEVKID NSQVESGSLE DDWDFLPPKK IKDPDAAKPE DWDERAKIDD PTDSKPEDWD KPEHIPDPDA KKPEDWDEEM DGEWEPPVIQ NPEYKGEWKP RQIDNPDYKG TWIHPEIDNP EYSPDANIYA YDSFAVLGLD LWQVKSGTIF DNFLITNDEA YAEEFGNETW GVTKAAEKQM KDKQDEEQRL KEEEEDKKRK EEEEAEDKED DDDRDEDEDE EDEKEEDEEE SPGQAKDEL | |
Greater than 90% as determined by SDS-PAGE. |
12317 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |