missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CCL3 / Mip1a Protein
A DNA sequence encoding the Mouse CCL3 / Mip1a, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10263-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_035467.1 | |
CCL3 / Mip1a Protein | |
Research Use Only | |
CCL3 | |
The biological activity determined by a chemoattract bioassay using murine splenocytes is in a concentration range of 10-100 ng/ml. | |
Mouse | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
20302 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APYGADTPTA CCFSYSRKIP RQFIVDYFET SSLCSQPGVI FLTKRNRQIC ADSKETWVQE YITDLELNA | |
Greater than 98% as determined by SDS-PAGE and HPLC. |