missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD14 Protein
A cDNA sequence encoding the CD14 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11301-HIS-INSECT-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
12475 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MERVLGLLLL LLVHASPAPP EPCELDEESC SCNFSDPKPD WSSAFNCLGA ADVELYGGGR SLEYLLKRVD TEADLGQFTD IIKSLSLKRL TVRAARIPSR ILFGALRVLG ISGLQELTLE NLEVTGTAPP PLLEATGPDL NILNLRNVSW ATRDAWLAEL QQWLKPGLKV LSIAQAHSLN FSCEQVRVFP ALSTLDLSDN PELGERGLIS ALCPLKFPTL QVLALRNAGM ETPSGVCSAL AAARVQLQGL DLSHNSLRDA AGAPSCDWPS QLNSLNLSFT GLKQVPKGLP AKLSVLDLSY NRLDRNPSPD ELPQVGNLSL KGNPFLDSES HSEKFNSGVV TAGAPSSQAV ALSGTLALLL GDRLFV |
CD14 Protein | |
Research Use Only | |
Cd14 | |
Mouse | |
His | |
CD14 was lyophilized from a concentrated protein solution (1 mg/ml) containing phosphate-buffered saline, pH 7.2. |