missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EGF / Epidermal Growth Factor Protein
Recombinant Protein
Marca: enQuireBio™ QP5482-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_034243.2 | |
EGF / Epidermal Growth Factor Protein | |
20 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
3T3 cell proliferation significantly induced by less than 250 pg/ml of Mouse EGF / Epidermal Growth Factor (which is equivalent to activity of at least 4.0 x 10^6 units/mg or an ED50 typically around 10-100 pg/ml). | |
Mouse | |
Biotin | |
0.5 mg/mL in sterile PBS |
13645 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
EGF | |
Recombinant Protein | |
E. coli | |
MKKIDDDKNS . YPGCPSSYD G. YCLNGGVC MH. IESLDSY TCN. CVIGYS GDRC. QTRDL EWWEL. R |