missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Egl nine homolog 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Egl nine homolog 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8813-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_082409.2 | |
Egl nine homolog 3 Protein | |
Research Use Only | |
Egln3 | |
Recombinant Protein | |
E. coli | |
PLGHIMRLDL EKIALEYIVP CLHEVGFCYL DNFLGEVVGD CVLERVKQLH YNGALRDGQL AGPRAGVSKR HLRGDQITWI GGNEEGCEAI NFLLSLIDRL VLYCGSRLGK YYVKERSKAM VACYPGNGTG YVRHVDNPNG DGRCITCIYY LNKNWDAKLH GGVLRIFPEG KSFVADVEPI FDRLLFFWSD RRNPHEVQPS YATRYAMTVW YFDAEERAEA KKKFRNLTRK TESALAKD | |
Greater than 90% as determined by SDS-PAGE. |
112407 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |