missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GSTA1 Protein
A cDNA sequence encoding the GSTA1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12056-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
GSTA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGKPVL HYFNARGRME CIRWLLAAAG VEFEEKFIQS PEDLEKLKKD GNLMFDQVPM VEIDGMKLAQ TRAILNYIAT KYDLYGKDMK ERALIDMYSE GILDLTEMIG QLVLCPPDQR EAKTALAKDR TKNRYLPAFE KVLKSHGQDY LVGNRLTRVD IHLLEVLLYV EEFDASLLTP FPLLKAFKSR ISSLPNVKKF LQPGSQRKPP MDAKQIQEAR KAFKIQ | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
GSTA1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |