missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HDAC8 Protein
A cDNA sequence encoding the HDAC8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12203-1ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
HDAC8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEMPEEPANS GHSLPPVYIY SPEYVSICDS LVKVPKRASM VHSLIEAYAL HKQMRIVKPK VASMEEMATF HTDAYLQHLQ KVSQEGDEDH PDSIEYGLGY DCPATEGIFD YAAAIGGGTI TAAQCLIDGK CKVAINWSGG WHHAKKDEAS GFCYLNDAVL GILRLRRKFD RILYVDLDLH HGDGVEDAFS FTSKVMTVSL HKFSPGFFPG TGDMSDVGLG KGRYYSVNVP IQDGIQDEKY YHICESVLKE VYQAFNPKAV VLQLGADTIA GDPMCSFNMT PVGIGKCLKY VLQWQLATLI LGGGGYNLAN TARCWTYLTG VILGKTLSSE IPDHEFFTAY GPDYVLEITP SCRPDRNEPH RIQQILNYIK GNLKHVVHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
HDAC8 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |