missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Hemoglobin subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Hemoglobin subunit alpha, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6146-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Hemoglobin subunit alpha Protein | |
Research Use Only | |
Hba | |
Recombinant Protein | |
E. coli | |
VLSGEDKSNI KAAWGKIGGH GAEYGAEALE RMFASFPTTK TYFPHFDVSH GSAQVKGHGK KVADALASAA GHLDDLPGAL SALSDLHAHK LRVDPVNFKL LSHCLLVTLA SHHPADFTPA VHASLDKFLA SVSTVLTSKY R | |
Greater than 80% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |