missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LBP Protein
A cDNA sequence encoding the LBP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10742-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
16803 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Up to 2microg/ml LBP mediates binding of FITC-LPS (0.5microg/ml) to CD14+CHO transfectants (2 x 106/ml). | |
Mouse | |
His | |
Recombinant Mouse LBP was lyophilized from a protein solution (1 mg/ml) containing phosphate-buffered saline, pH 7.2. |
LBP Protein | |
Research Use Only | |
Lbp | |
Recombinant Protein | |
Mouse | |
MKAGTGPLLS TLLGLLFLSI QGTGGVNPGV VARITDKGLA YAAKEGLVAL QRELYKITLP DFSGDFKIKA VGRGQYEFHS LEIQNCELRG SSLKLLPGQG LSLAISDSSI GVRGKWKVRK SFLKLHGSFD LDVKGVTISV DLLLGMDPSG RPTVSASGCS SRICDLDVHI SGNVGWLLNL FHNQIESKLQ KVLENKVCEM IQKSVTSDLQ PYLQTLPVTA EIDNVLGIDY SLVAAPQAKA QVLDVMFKGE IFNRNHRSPV ATPTPTMSLP EDSKQMVYFA ISDYAFNIAS RVYHQAGYLN FSITDDMLPH DSGIRLNTKA FRPFTPQIYK KYPDMKLELL GTVVSAPILN VSPGNLSLAP QMEIEGFVIL PTSAREPVFR LGVVTNVFAS LTFNNSKVTG MLHPDKAQVR LIESKVGMFN VNLFQAFLNY YLLNSLYPDV NAELAQGFPL PLPRHIQLHD LDFQIRKDFL YLGANVQYMR V |