missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MAP3K1 Protein
A DNA sequence encoding the Mus musculus (Mouse) MAP3K1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8830-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005912.1 | |
MAP3K1 Protein | |
Research Use Only | |
MAP3K1 | |
Recombinant Protein | |
E. coli | |
IIIIQQDTPE TLPGHTKAKQ PYREDTEWLK GQQIGLGAFS SCYQAQDVGT GTLMAVKQVT YVRNTSSEQE EVVEALREEI RMMSHLNHPN IIRMLGATCE KSNYNLFIEW MAGGSVAHLL SKYGAFKESV VINYTEQLLR GLSYLHENQI IHRDVKGANL LIDSTGQRLR IADFGAAARL ASKGTGAGEF QGQLLGTIAF MAPEVLRGQQ YGRSCDVWSV GCAIIEMACA KPPWNAEKHS NHLALIFKIA SATTAPSIPS HLSPGLRDVA LRCLELQP | |
Greater than 90% as determined by SDS-PAGE. |
4214 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |